Imprimer
Centrifuge tubes, 15 ml, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
430052 430053 430055 430766 430790 430791 431470
734-1812EA 180 EUR
734-1812 734-1813 734-1814 734-1862 734-1866 734-1867 525-0477
Centrifuge tubes, 15 ml, Corning®
Tubes Tubes à centrifuger
Corning® 15 ml clear centrifuge tubes are provided without caps for ease of use in automated or other applications where non pre-attached caps are desired.
  • Black printed graduations and a large white marking spot
  • Maximum RCF is 12000×g
  • Sterile, certified nonpyrogenic and DNase-/RNase-free
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping. This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Informations de commande: Foam racks and CentriStar™ caps can be purchased separately.
Cdt: Bulk packed 25 per resealable zip lock bag.