Imprimer
Amine and Carboxyl Plates, PureCoat™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
354721 354723 356721 356723 356775 354775
734-1471EA 73 EUR
734-1471 734-1473 734-1472 734-1474 734-1486 734-1485
Amine and Carboxyl Plates, PureCoat™
Microplaques Microplaques pour culture cellulaire
Corning PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces.
These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.