Imprimer
Culture inserts, BRAND Insert 2in1, for 6-well plates, sterile, BIO-CERT® cell culture quality
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
782752 782753 782754 782755
734-3683EA 267 EUR
734-3683 734-3684 734-3685 734-3686
Culture inserts, BRAND Insert 2in1, for 6-well plates, sterile, BIO-CERT® cell culture quality
Microplaques Inserts pour plaques de cultures
The BRAND Insert 2in1 is a cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more.
  • Inserts can be used in a hanging or standing position
  • For 24, 12, and 6-well plates or culture dishes
  • Achieve uniform and controlled cell growth thanks to surface treatment
  • Suitable for 2D and 3D cell cultures
In many cell culture tasks, or when switching from a standing to a hanging position, you can continue to use your current cell culture plates, saving additional validation steps. Thanks to both supporting feet and hooks, you can position the BRAND Insert 2in1 either way. Via its protein-like structure, the cellGrade™ plus surface provides optimal conditions for growing primary keratinocytes and many other cell types. With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row, so that the remaining inserts stay sterile. Besides the practical benefits, this also reduces waste.
Certifications: Sterility Assurance Level (SAL) of 10⁻⁶.
Cdt: Packaged individually or in practical multi-packs.