Imprimer
Stainless steel trolley with shelves
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BC0011/3 BC0007/3
TEKMBC0011/3EA 0 EUR
TEKMBC0011/3 TEKMBC0007/3
Stainless steel trolley with shelves
Chariots
Our multi-tier general purpose trolley is manufactured from fully welded 304-grade stainless steel, making it robust, long-lasting and hygienic.
  • Fully welded 304-grade stainless steel
  • Brushed dull polish finish
  • Wall bump guards
  • Swivel castors with brake
Our multi-tier general purpose trolley is manufactured from fully welded 304-grade stainless steel in a brushed dull polish finish, making it not only robust but extremely intelligent too; from quiet wheels to shelves with sound-absorbing lining and a 200 kg payload per shelf.
It is the ideal hygienic solution to your transportation and storage needs, being easy to clean and versatile, the perfect trolley for your workplace. The build and stainless-steel quality also available in 316-grade which means it is extremely long-lasting and is guaranteed to be corrosion resistant.
As with all our products, our multi-tier general purpose trollies are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workplace.
The trolley comes complete with 4 quiet castors, 2 of which are braked as standard for ultimate versatility and stability. It also features wall bump guards, enabling you to maintain a safe and hazard-free workspace.
Informations de commande: Narrow version with inset castors available. For maximum security, anti-static or rubber castors are also available. For more information, contact your Avantor representative.