Imprimer
MyPIPETMAN® Select Pipettors, Gilson
Fournisseur: GILSON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FP10012S FP10014S FP10001S FP10002S FP10003S FP10004S FP10006S FP10007S FP10008S FP10009S FP10010S FP10011S FP10013S FP10015S FP10016S
613-7020EA 1143 EUR
613-7020 613-7022 613-7009 613-7010 613-7011 613-7012 613-7014 613-7015 613-7016 613-7017 613-7018 613-7019 613-7021 613-7023 613-7024
MyPIPETMAN® Select Pipettors, Gilson
Pipettes Pipetteurs
Ergonomic, safe, and reliable, MyPIPETMAN® Select offers volume setting choice and optional handle personalisation for optimal use in various protocols.
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
Ergonomic design
Lightweight and well-balanced, MyPIPETMAN® Select has an elegant design with a textured effect for a better grip. With no compromise on quality and performance, the built-in ejection system with an integrated lever provides reduced tip ejection forces compared to direct ejection systems. This ejection system reinforces the pipette's ergonomics for convenient use throughout the day.
Personalise your pipette for an optimal use
MyPIPETMAN® Select has optional handle printing on demand. Identify your pipette to avoid any risk of contamination by dedicating it to a specific liquid or workflow step and improve your lab's Lean Management process.