Imprimer
Manganèse (II) chlorure tétrahydraté ≥98% FCC
Marques : Spectrum Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
M1109-2.5KG M1109-25KGBL M1109-125GM M1109-500GM
SPCMM1109-2.5KGEA 936 EUR
SPCMM1109-2.5KG SPCMM1109-25KGBL SPCMM1109-125GM SPCMM1109-500GM
Manganèse (II) chlorure tétrahydraté ≥98% FCC
Manganèse (II) chlorure tétrahydraté
Manganese Chloride, Tetrahydrate, FCC is used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: MnCl₂·4H₂O
Point d'ébullition: 1190 °C (1013 hPa)
Point de fusion: 58…59 °C
Densité: 2,01 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00149792
Numéro CAS: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Indice Merck: 13,05751
Commander maintenant
Résultats des tests de spécification
Assay 98% Min.