Imprimer
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Marques : Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
X193-04EA 1679 EUR
X193-04 X193-01
J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Tampons Tampons d'extraction
J.T. Baker® Cell Lysis Solution. Optimized multi-component, ready-to-use solution for extraction and recovery of AAV particles from HEK293 or SF9 cells. The product offers quantitative recovery through efficient cell lysis and protection of vector particles against shear stress. The solution is formulated with non-ionic detergents which are octyl phenol ethoxylate free.
  • Comparable cell lysis performance to octyl phenol ethoxylate
  • Improved protection of viral particles
  • Reduced foaming and viscosity
  • 0.2 µm filtered solution
Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)
Chemical Specifications of Non-GMP Cell lysis solution:
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total microbial count: No growth detected
UV-Vis (max Absorption) at 380-450 nm: For information only