Imprimer
Lumitester smart, Kikkoman
Fournisseur: HYSERVE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1702671 1702672 1902653
710-2244EA 375 EUR
710-2244 710-2245 710-2243
Lumitester smart, Kikkoman
Systèmes de surveillance Tests de contrôle de l'hygiène
Luminometer for hygiene validation of surfaces in 10 seconds.
The lumitester Smart, in combination with the LuciPac A3 reagent tests, detects all organic residues and microorganisms on surfaces as well as in liquids and determines the degree of cleanliness at the point of sampling. The system thus enables hygiene monitoring of surfaces within seconds, directly following cleaning and disinfection measures.
The Lumitester Smart is a mobile device that allows it to be used even in places that are difficult to access, making it particularly user-friendly. Via Bluetooth, the device can be connected to smartphones, tablets, and laptops, where comprehensive evaluation of the measurement data is possible. By storing the data in the cloud, the results from multiple users and devices can be combined and evaluated regardless of location.
Informations de livraison: It includes:1 lumitester smart,1 wrist strap, 1 USB cable, 2 batteries (AA, Mignon), 3 cleaning brushes and a quick start guide.