Imprimer
Lecteur HH83 UHF ACD 2Dimg EU
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
310380 310381
BRDY310380EA 2550 EUR
BRDY310380 BRDY310381
Lecteur HH83 UHF ACD 2Dimg EU
Systèmes de code-barres
The HH83 UHF ACD scanners are an excellent RFID scanning choice and are a perfect match for Brady SmartID solutions. Available in two versions: With or without LTE.
  • Reads all major 1D barcodes and 2D codes
  • Rugged scanners for data collection in challenging industrial environments
  • Ergonomic design for extended usage IP65 rating and fully drop-tested
  • Perfect RFID legibility for a multitude of RFID labels
  • Extended read range by using di-pole antenna
  • Visual (LED’s) and haptic (vibration) feedback
  • Battery Hotswap: Replace an empty battery and install a fully charged one in a matter of seconds
The Brady HH83 models are perfect for daily RFID scanning. They enable users to pick up a large range of RFID signals and offer a great user experience.
RFID Capabilities: NFC/HF and UHF
Reading Speed: Up to 1000 tags per second
Focal Point: Up to 15 m
Display Resolution: 1280 x 720 pixels
Connectivity Options:
Non-LTE model: Bluetooth, USB 2.0, Wireless 802.11 a/b/g/n/ac
LTE model: 3G, 4G, Bluetooth, GPRS/EDGE, USB 2.0, Wireless 802.11 a/b/g/n/ac
Informations de livraison: Supplied with changeable battery (pre-installed), safety and regulations card, wrist strap.