Imprimer
GOJO® Antimicrobial Plus Foam Handwash Refills
Fournisseur: GOJO Industries - Europe Ltd
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5348-02-EEU00 5148-03-EEU00
GOJO5348-02-EEU00EA 71.3 EUR
GOJO5348-02-EEU00 GOJO5148-03-EEU00
GOJO® Antimicrobial Plus Foam Handwash Refills
Soin de la peau Savons et soins nettoyants
La mousse lavante antimicrobienne GOJO® sans quat est douce pour la peau et formulée pour être très efficace.
  • Suitable for sensitive skin and formulated for high frequency use
  • Enriched with a moisturising plant-based agent
  • Broad Spectrum efficacy (bacteria, yeasts, and virus)
  • Passes EN 1499 in 30 seconds
  • Targets food specific pathogens such as Salmonella, Listeria, and E. coli
  • Unfragranced and colourless
  • GOJO CLEAN SEALED™ refills snap quickly and easily into place; protects users from the contamination risks of bulk soap
  • Refills made with 100% R-PET
  • Halal certified
Convient aux zones de manipulation des aliments telles que les restaurants, les bureaux d'accueil et de service, les cuisines, la transformation des aliments et la gestion des installations.
Certifications: Conforme au test d'altération des aliments : BS EN ISO 4120:2007.
Informations de commande: Some of these refills are part of the GOJO free dispenser program. To receive a FREE of CHARGE manual dispenser, you need to purchase at least one refill. The manual dispensers available for this offer are GOJO® ADX-12™, GOJO® ADX-7™, GOJO® PRO TDX™ and GOJO® FMX™. If you want a FREE of CHARGE touch-free dispenser, you need to purchase one case consisting of two refills to receive a GOJO® TFX™ white and two cases of two refills each to receive a GOJO® TFX™ metallic.