Imprimer
Semi-micro laboratory balances, Cubis® II MCE
Fournisseur: Sartorius Balances
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MCE225P2S00-U MCE225S2S00-U MCE125S2S00-U MCE125P2S00-U MCE225S2S00-A MCE225P2S00-A MCE125S2S00-A MCE125P2S00-A MCE225S2S00-I MCE225P2S00-I MCE125S2S00-I MCE125P2S00-I
SARBMCE225P2S00-UEA 12390 EUR
SARBMCE225P2S00-U SARBMCE225S2S00-U SARBMCE125S2S00-U SARBMCE125P2S00-U SARBMCE225S2S00-A SARBMCE225P2S00-A SARBMCE125S2S00-A SARBMCE125P2S00-A SARBMCE225S2S00-I SARBMCE225P2S00-I SARBMCE125S2S00-I SARBMCE125P2S00-I
Semi-micro laboratory balances, Cubis® II MCE
Balances and Scales
Cubis® II MCE semi-micro balances with essential user interface offer readability of 10 µg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the µg-range.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status centre notifications
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis® II balance series from Sartorius.
All weighing modules of the Cubis® II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
These MCE models with essential user interface have a black and white touch screen plus holders for ergonomic sample handling and automatic motorised levelling. The balances feature innovative draft shield technology, gesture sensor and climate monitoring. With complete traceability the balances help achieve 21 CFR Part 11 and EU Annex 11 compliance.
Choice of draft shield denoted by model name suffix: manual glass doors with large draft shield (U), motorised draft shield (A) or motorised with built-in de-ioniser (I) for effective removal of static charges.