Imprimer
Serological pipettes, polystyrene, sterile, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
56800-10110 56800-1110 56800-25210 56800-50510
WHEA56800-10110EA 174 EUR
WHEA56800-10110 WHEA56800-1110 WHEA56800-25210 WHEA56800-50510
Serological pipettes, polystyrene, sterile, WHEATON®
Pipettes Pipettes sérologiques
WHEATON® serological pipettes provide the performance required to succeed at every stage of cell culture and general life science research.
  • Manufactured from polystyrene compliant with USP Class VI
  • Range 1 to 50 ml, ideal for cell culture and general laboratory applications
  • Calibrated to deliver (TD) to ±2%
  • Plugged and supplied sterile by gamma irradiation
  • Non-cytotoxic and certified non-pyrogenic to <0,5 EU/ml
Clear easy to read bi-directional graduations give the choice between ascending and descending scale, with negative graduations for extra capacity.
Informations sur les accessoires: The Wheaton Pipette Rack (WHEA72190-0000) is ideal to hold serological pipettes and associated pipette controllers.
Cdt: Available in 3 convenient packaging options to suit most user needs:
- 56900 series are supplied individually wrapped in paper plastic peel packaging
- 56800 series supplied bulk packed in plastic bags
- 56700 series are packed in a cardboard canister