Imprimer
Qualitative filter papers, Grade 598 ½, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10312244 10312247 10312251 10312256
515-5204EA 104 EUR
515-5204 512-3635 515-5207 515-5211
Qualitative filter papers, Grade 598 ½, Whatman™
Filtres Papiers filtre Papiers filtres qualitatifs
A thick prepleated filter paper with high loading capacity. Combines medium retention with medium-fast to quick filtration speed.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.