Imprimer
Caps for Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PBCC20-ST3 6PBCC20-ST3F 6PBCC20-ST3T 6PBCC20-STH 6PBCC20-STHT 6PBCC20-STME 6PBCC20-STMEF 6PBCC20-STMET 6PCC20-ST3 6PCC20-ST3F 6PCC20-ST3T
HYPE6PBCC20-ST3EA 89.2 EUR
HYPE6PBCC20-ST3 HYPE6PBCC20-ST3F HYPE6PBCC20-ST3T HYPE6PBCC20-STH HYPE6PBCC20-STHT HYPE6PBCC20-STME HYPE6PBCC20-STMEF HYPE6PBCC20-STMET HYPE6PCC20-ST3 HYPE6PCC20-ST3F HYPE6PCC20-ST3T
Caps for Glass Crimp Top Headspace Vials, Level 3 High Performance Applications, SureSTART™
Capuchons Bouchons pour flacons de chromatographie
Use Thermo Scientific™ SureSTART™ glass crimp top headspace vials, performance level 3, for your volatile gas analyses.
Available in 6, 10 and 20 ml clear glass formats, these headspace vials are made to withstand higher temperatures and internal pressures required for volatile gas analyses. This is a performance level 3 vials, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. They are specification certified to ensure physical dimensions and autosampler compatibility. These vials meet all requirements of US, EU, JPN Pharmacopeia. Use 20 mm crimp caps and septa to seal these headspace vials. Buy in bulk for greater savings.