Imprimer
Total recovery glass screw top 1,5 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PSV9-TR1 6PSV9-TR1F 6PSV9-TR1T 6PSV9-CHV1 6PSV9-SV1
HYPE6PSV9-TR1EA 386 EUR
HYPE6PSV9-TR1 HYPE6PSV9-TR1F HYPE6PSV9-TR1T HYPE6PSV9-CHV1 HYPE6PSV9-SV1
Total recovery glass screw top 1,5 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Fioles
Choose Thermo Scientific™ SureSTART™ 1,5 ml total recovery glass screw top microvials, performance Level 3, when you need to maximise the injection volume for <2 ml samples.
Available in clear and amber glass, this one-piece total recovery microvials has a very low residual volume due to a specially-shaped deep and narrow inner reservoir which collects the liquid in the base so that compatible autosamplers can collect all but 1-2 µl of sample while having a total sample volume of 1,2 ml and a residual volume of 1 µl. The outer dimensions for these vials are 12×32 mm. These performance Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.