Imprimer
Glass screw top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PSV9-03FIVP 6PSV9-03FIVAP 6PSV9-03FIVAPF 6PSV9-03FIVAPT 6PSV9-03FIVPF 6PSV9-03FIVPT
548-2771EA 82.96 EUR
548-2771 HYPE6PSV9-03FIVAP HYPE6PSV9-03FIVAPF HYPE6PSV9-03FIVAPT HYPE6PSV9-03FIVPF HYPE6PSV9-03FIVPT
Glass screw top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
Fioles
Choose Thermo Scientific™ SureSTART™ 0,3 ml glass screw top microvials, performance level 3, when you need to maximise the injection volume for <2 ml samples.
This 12×32 mm microvials has a fixed insert that provides a total usable volume of 0,25 ml and a residual volume of 4 µl. Available in clear and amber glass, this is a performance level 3 vial, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.