Imprimer
Glass screw top vials, 2 ml, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PSV9-1P 6PSV9-1PF 6PSV9-1PSS 6PSV9-1PSSF 6PSV9-1PSST 6PSV9-1PT 6PSV9-2PSS 6PSV9-2PSSF 6PSV9-2PSST 6PSV9-S1P
HYPE6PSV9-1PEA 0 EUR
HYPE6PSV9-1P HYPE6PSV9-1PF 548-2773 HYPE6PSV9-1PSSF HYPE6PSV9-1PSST HYPE6PSV9-1PT HYPE6PSV9-2PSS HYPE6PSV9-2PSSF HYPE6PSV9-2PSST HYPE6PSV9-S1P
Glass screw top vials, 2 ml, level 3 high performance applications, SureSTART™
Fioles
Choose Thermo Scientific™ SureSTART™ 2 ml GOLD-grade clear glass screw and crimp top vials, performance level 3, when analysing analytes such as polar compounds that strongly adsorb on the glass surface or are sensitive to sample pH changes.
Level 3 vials are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. This offering includes Thermo Scientific™ SureStop™ vials, which, when used with 9 mm AVCS™ screw caps, ensure an optimal seal every time. The SureStop vial-AVCS cap system is specially designed to prevent screw thread vial failures such as cap tilt, cap deformities, dislodged septa, or septum push through, thus preventing injection failures and sample evaporation.