Imprimer
Polypropylene snap top 2 ml microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6ERV11-03PPC 6ERV11-03PPCF 6ERV11-03PPCT 6ERV11-08PPC 6ERV11-08PPCF 6ERV11-08PPCT 6ERV11-08PPFB 6ERV11-08PPFBF 6ERV11-08PPFBT
HYPE6ERV11-03PPCEA 27.4 EUR
HYPE6ERV11-03PPC HYPE6ERV11-03PPCF HYPE6ERV11-03PPCT HYPE6ERV11-08PPC HYPE6ERV11-08PPCF HYPE6ERV11-08PPCT HYPE6ERV11-08PPFB HYPE6ERV11-08PPFBF HYPE6ERV11-08PPFBT
Polypropylene snap top 2 ml microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Fioles
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml polypropylene snap top microvials for everyday chromatography analysis of <2 ml samples. They provide an affordable choice and can be used with all HPLC/UHPLC instrument types.
These clear plastic 2 ml snap top vials are specification certified to ensure autosampler compatible physical dimensions. Seal these snap top vials with 11 mm snap caps and septa.