Imprimer
Polypropylene screw top 2 ml microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6ESV9-04PP 6ESV9-04PPF 6ESV9-04PPT 6ESV9-1PP 6ESV9-1PPF 6ESV9-1PPT
HYPE6ESV9-04PPEA 27.5 EUR
HYPE6ESV9-04PP HYPE6ESV9-04PPF HYPE6ESV9-04PPT HYPE6ESV9-1PP HYPE6ESV9-1PPF HYPE6ESV9-1PPT
Polypropylene screw top 2 ml microvials for <2 ml samples, level 1 everyday analysis, SureSTART™
Fioles
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml polypropylene screw top microvials for everyday chromatography analysis of <2 ml samples. They provide an affordable choice and can be used with all HPLC/UHPLC instrument types.
These clear plastic 2 ml screw top microvials are specification certified to ensure autosampler compatible physical dimensions. Seal these screw top vials with 9 mm screw caps and septa.