Imprimer
Glass crimp top vials, 2 ml, level 1 everyday analysis, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CHCV11-10P CHCV11-10PF CHCV11-10PT CHCV11-20P CHCV11-20PF CHCV11-20PT
HYPECHCV11-10PEA 14.4 EUR
HYPECHCV11-10P HYPECHCV11-10PF HYPECHCV11-10PT HYPECHCV11-20P HYPECHCV11-20PF HYPECHCV11-20PT
Glass crimp top vials, 2 ml, level 1 everyday analysis, SureSTART™
Fioles
Use performance level 1 Thermo Scientific™ SureSTART™ 2 ml glass crimp top vials for everyday chromatography analysis. They provide an affordable choice and can be used with all GC and HPLC/UHPLC instrument types.
These 2 ml glass crimp top vials are specification certified to ensure autosampler compatible physical dimensions. Choose from clear and amber glass. The vials meet all requirements of US, EU, and JPN pharmacopia. Seal these crimp vials with 11 mm aluminium crimp caps and septa using a manual or electronic crimper tool.