Imprimer
Glass high recovery screw top 1,7 ml microvials for <2 ml samples, level 2 high-throughput applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6ASV9-V1 6ASV9-V1T
HYPE6ASV9-V1EA 216 EUR
HYPE6ASV9-V1 HYPE6ASV9-V1T
Glass high recovery screw top 1,7 ml microvials for <2 ml samples, level 2 high-throughput applications, SureSTART™
Fioles
Choose Thermo Scientific™ SureSTART™ 1,7 ml high recovery glass screw top microvials to maximise the injection volume for <2 ml samples in high-throughput applications.
With dimensions of 12×32 mm, these microvials have an internal conical reservoir that provides a total usable volume of 1,5 ml and a residual volume of 4 µl. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. Use level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. They have very low metal content, extractables, and leachables and conform to high standards to meet your regulatory requirements. These vials are specification certified to ensure their physical dimensions and autosampler compatibility.