Imprimer
Specification certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PK1655 6PKG592W
HYPE6PK1655EA 113 EUR
HYPE6PK1655 HYPE6PKG592W
Specification certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Fioles
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits.
Select Level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Choose from a variety of kits containing short thread screw vials in glass or TPX and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.