Imprimer
HPLC and GC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PCK575W 6PCK576W 6PCK578W 6PCK593W 6PCK76W 6PCK93W
HYPE6PCK575WEA 79.1 EUR
HYPE6PCK575W HYPE6PCK576W HYPE6PCK578W HYPE6PCK593W 548-2766 548-2767
HPLC and GC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Fioles
Meet your certification and documentation needs with our level 3 Thermo Scientific™ SureSTART™ HPLC and GC certified screw vial and cap kits. Select level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
Choose from a variety of kits containing short thread screw vials with marking spots in clear or amber glass and polypropylene caps with silicone/PTFE septa designed to reduce autosampler needle issues. Pre-slit and bonded septa options are available. Select kits with SureStop™ vials and/or AVCS™ screw caps for effective seals, consistent data, and increased sample throughput.