Imprimer
Total recovery glass snap top microvials, 1,5 ml for <2 ml samples, level 3 high performance applications, SureSTART™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6PRV11-TR1 6PRV11-TR1T
HYPE6PRV11-TR1EA 309 EUR
HYPE6PRV11-TR1 HYPE6PRV11-TR1T
Total recovery glass snap top microvials, 1,5 ml for <2 ml samples, level 3 high performance applications, SureSTART™
Fioles
Choose Thermo Scientific™ 1,5 ml total recovery glass snap top microvials, performance level 3, when you need to maximise the injection volume for <2 ml samples.
This one-piece, 12×32 mm total recovery microvial has a very low residual volume due to a specially shaped deep and narrow inner reservoir. This configuration collects the liquid in the base so that compatible autosamplers can inject all but 1 to 2 µl of sample while having a total sample volume of 1,2 ml and a residual volume of 1 µl. These are performance level 3 vials, which means they are designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.