Imprimer
Floor mattings, Notrax® 827 Anti-Stat P.O.P.™
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
827R3040BU 827R3640BU 827C0024BU 827C0030BU 827C0036BU
115-2898EA 686 EUR
115-2898 115-2899 115-2901 115-2902 115-2903
Floor mattings, Notrax® 827 Anti-Stat P.O.P.™
Floor Mats Tacky Mats
Anti-Stat P.O.P.™ is a vinyl work surface that dissipates static from conductive items placed on it.
  • Smooth top surface
  • ESD static dissipative
  • Meets EOS/ESD-S4, measured resistance for work surfaces
  • Electrostatic charge (walking test), meets ISO6356 and EN 1815
  • Grounding cord available, not required
Protect and keep sensitive components safe from damaging static shocks. Smooth top surface is abrasion resistant and easy to wipe clean.
Material Sponge/ESD.