Imprimer
Novagen® GST Bind™ Resin, MilliporeSigma
Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
70541-3 70541-4 70541-5
70541-3EA 461 EUR
70541-3 70541-4 70541-5
Novagen® GST Bind™ Resin, MilliporeSigma
Protéines purification Protein Purification Resins
GST Bind™ Resin utilizes an 14-atom spacer arm to covalently attach reduced glutathione via a sulfide linkage. The high degree of substitution of glutathione ensures a high binding capacity with yields of glutathione S-transferase (GST) fusion proteins of 5 to 8 mg/ml settled resin. The resin can be re-used up to 6 times without loss of capacity.