Imprimer
Stainless steel lids for water baths
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SL4-8 SL4-14 SL4-22
462-0067EA 552 EUR
462-0067 462-0065 462-0066
Stainless steel lids for water baths
Bains Bains-marie à circulation
Stainless steel gable lids helps reduce evaporation, assists in keeping samples in a water bath free from contamination and saves energy.
  • High profile allows extended use of the bath, right into the edges of the tank
  • No drip points to contaminate samples whilst the lid design ensures all condensate returns back into the bath
  • Insulated handles for safe use