Imprimer
Rotor package, Avanti JCF-Z
Fournisseur: Beckman Coulter
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
335140 357521
521-3596EA 0 EUR
521-3596 521-3912
Rotor package, Avanti JCF-Z
Centrifugeuses Rotors
Particles sediment out of the flowing stream, which then emerges from the rotor as a particle-depleted effluent. This process continues until the particle containing capacity of the rotor is reached, or until the starting material (sample) is completely processed. Thus the amount of sample that can be processed in one run is determined by the sample’s volume and concentration of particles.