Imprimer
Screw cap microcentrifuge tubes, SuperClear®
Fournisseur: LABCON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3610-870-000-9 3611-870-000-9 3611-875-000-9 3613-870-000-9 3613-875-000-9 3620-870-000-9 3621-870-000-9 3621-875-000-9 3623-870-000-9 3623-875-000-9 3633-870-000-9 3633-875-000-9 3643-870-000-9 3643-875-000-9 3650-870-000-9 3651-870-000-9 3651-875-000-9 3653-870-000-9 3653-875-000-9 3660-870-000-9 3661-870-000-9 3661-875-000-9 3663-870-000-9 3663-875-000-9 3641-875-000-9 3640-870-000-9
3610-870-000-9EA 93.9 EUR
3610-870-000-9 3611-870-000-9 3611-875-000-9 3613-870-000-9 3613-875-000-9 3620-870-000-9 3621-870-000-9 3621-875-000-9 3623-870-000-9 3623-875-000-9 3633-870-000-9 3633-875-000-9 3643-870-000-9 3643-875-000-9 3650-870-000-9 3651-870-000-9 3651-875-000-9 3653-870-000-9 3653-875-000-9 3660-870-000-9 3661-870-000-9 3661-875-000-9 3663-870-000-9 3663-875-000-9 3641-875-000-9 3640-870-000-9
Screw cap microcentrifuge tubes, SuperClear®
Tubes Microtubes à centrifuger
SuperClear® tubes are made of high quality medical grade ultra clear resins to ensure your work is safe and repeatable. These tubes provide a clear view of the tube contents as well as correct colour rendition and fit all standard and high capacity microcentrifuge rotors
  • Tubes are available freestanding or with a conical bottom
  • ATP, RNase/DNase and endotoxin free (non-pyrogenic)
  • Tubes and caps are autoclavable
  • Can be centrifuged to 20000 RCF
Available options:
- Tubes with O-ring caps (ethylene propylene)
- Tubes with elastomeric sealing ring
- Tubes only
- Caps only
Cdt: Packaged in resealable bags. Sterile versions of these tubes are packaged in medical grade bags. Sterilized with a validated medical grade process to an SAL of 10⁻⁶.