Imprimer
Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices
Fournisseur: Oxford Nanopore Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ADP-FLG001 FLO-FLG114.6 FLG.SP
ADP-FLG001EA 895 EUR
ADP-FLG001 FLO-FLG114.6 OXNTFLG.SP
Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices
Cuves à circulation
Flongle is an adapter for MinION and GridION devices that enables direct, real-time DNA sequencing on smaller, single-use Flongle Flow Cells.
  • The most rapid, accessible, and cost-efficient nanopore sequencing system for smaller or more frequently performed assays and experiments
  • Each Flongle Flow can deliver 1–2 Gb of real-time sequencing data
The Flongle Adapter allows MinION and GridION devices to run smaller, single-use Flongle Flow Cells, typically generating 1–2 Gb of data per flow cell. Users can simply insert a Flongle Flow Cell into the Flongle Adapter, run their experiment on their MinION or GridION device and then discard the Flongle Flow Cell and replace it with another.
How flow cells work: The user adds their prepared library to the flow cell and starts their sequencing experiment. Data is accumulated over time as DNA is sequenced by the nanopores. Individual read files are generated and are immediately available for analysis.
As data is streamed in real-time, a user can choose to stop a run as soon as sufficient data is gathered. A flow cell contains sufficient buffer to run for ~24 hours (under optimal conditions).
Flongle R10.4.1 Flow Cells have been tested with the following sequencing kits:
Ligation Sequencing Kit V14 (SQK-LSK114)
Ligation Sequencing Kit XL V14 (SQK-LSK114-XL)
Rapid Sequencing Kit V14 (SQK-RAD114)
Rapid Barcoding Kit 24 V14 (SQK-RBK114.24)
Rapid Barcoding Kit 96 V14 (SQK-RBK114.96)
Rapid PCR Barcoding Kit 24 V14 (SQK-RPB114.24)
Native Barcoding Kit 24 V14 (SQK-NBD114.24)
Native Barcoding Kit 96 V14 (SQK-NBD114.96)
16S Barcoding Kit 24 V14 (SQK-16S114.24)
Informations de livraison: The Flongle Adapter is stored and shipped at ambient temperature (15 to 25 °C).
Flow cells and kits are shipped together at 2 to 8 °C. The packaging is designed to protect the flow cells from freezing.
Flongle Flow Cells can be stored, unopened, at 2 to 8 °C for four weeks.
Cdt: Flongle flow cells are packed individually.
Attention: Not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Commander maintenant
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.