Imprimer
Masterflex® Single-Use Bag Port Disc Mini Dome, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX40331-00EA 39.2 EUR
MFLX40331-00 MFLX40331-01 MFLX40331-02 MFLX40331-03 MFLX40331-04 MFLX40331-06 MFLX40331-07 MFLX40331-08 MFLX40331-09 MFLX40331-10
Masterflex® Single-Use Bag Port Disc Mini Dome, Avantor®
Fittings and Connectors
Mini-dome protrusions inhibit suction blockage.
  • Barb sizes range from 1/8" to 1"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets.
If material validation is needed, please reach out to our techincal support representatives.