Imprimer
TCEP-HCl (Tris(2-carboxyéthyl)phosphine hydrochlorure), grade Premium, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): TCEP HCl
PG82080 PG82089 PG82090
PIERPG82080EA 104 EUR
PIERPG82080 PIERPG82089 PIERPG82090
TCEP-HCl (Tris(2-carboxyéthyl)phosphine hydrochlorure), grade Premium, Pierce™
TCEP-HCl (Tris(2-carboxyéthyl)phosphine hydrochlorure)
Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applications where product integrity and risk minimization are paramount.
  • TCEP—a thiol-free, odorless yet potent reducing agent for cleaving disulfide bonds
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade TCEP-HCl provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable. TCEP selectively and completely reduces even the most stable water-soluble alkyl disulfides over a wide pH range.
Attention: For Research Use Only. Not for use in diagnostic procedures.
Formule: C₉H₁₆ClO₆P
Poids moléculaire: 286.65 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00145469
Numéro CAS: 51805-45-9
UN: 3261
ADR: 8 II