Imprimer
Accupod solid samplers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8360T-10 8360T-10S 8360T-50 8360T-50S
300-0585EA 273 EUR
300-0585 300-0586 300-0587 300-0588
Accupod solid samplers
Échantillonneurs Préleveurs de solides
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
  • Suitable for sampling blend uniformity
  • Next generation in sampling technology
  • Designed to minimise sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.