Imprimer
Scoops, disposable, FoodScoop, SteriWare®
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8045B-1000 8045B-1000S 8045B-2500 8045B-500
SAMP8045B-1000EA 244 EUR
SAMP8045B-1000 SAMP8045B-1000S SAMP8045B-2500 SAMP8045B-500
Scoops, disposable, FoodScoop, SteriWare®
Pelles
These blue food scoops are ideal for use in the food industry.
  • Made from food grade Polypropylene
  • Available sterile and non sterile
  • Highly visible colour
  • Wide range of sizes
These blue food scoops can be easily spotted (if left behind in the powder) and are widely used throughout the food industry. They are both manufactured and packed in a cleanroom and are made from food grade polypropylene in line with HACCP guidelines.