Imprimer
Vacuum pump systems, remote-controlled, type SC 820 G and SC 840 G, LABOPORT®
Fournisseur: KNF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SC820G SC840G
181-0648EA 6580 EUR
181-0648 181-0649
Vacuum pump systems, remote-controlled, type SC 820 G and SC 840 G, LABOPORT®
Pompe Pompes à vide
Chemically resistant vacuum pump system comprising chemically resistant diaphragm vacuum pump, base plate, separator on suction side, high performance condenser on pressure side and vacuum controller.
  • Flow rate of up to 2,04 m³/h
  • Ultimate vacuum of 6 mbar abs
  • Remote-controlled for safe operation from outside closed fume hoods
  • High level of compatibility with vapor and condensation
  • Integrated gas ballast valve
  • Three color status display: In operation / Stand-by / Error
  • 100% oil-free transfer to ensure uncontaminated transfer, evacuation and compression
  • Equipped with an easy to clean surface
Ideal for rotary evaporation and other applications where control of vacuum level is important.
Pump head material: TFM™ PTFE
Diaphragm material: PTFE coated
Valve material: FFPM
Combination of TFM™ PTFE pump head and PTFE coated diaphragm makes these pumps ideal for working with extremely aggressive/corrosive gases and vapours.
Certifications: CE-Certification. ATEX compliant to (Ex) II 3/-G Ex h IIB+H2 T3 Gc internal atmosphere only.
Informations de livraison: Worldwide frequency and global plugs are included.