Imprimer
Compact burettes, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
13968 13963 13966
612-7164EA 110 EUR
612-7164 612-7162 612-7163
Compact burettes, BLAUBRAND®
Burettes
Borosilicate 3.3 glass, amber, class AS.
  • Removable PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • Calibrated to deliver (TD, Ex)
Certifications: DIN EN ISO 385, DE-M marking
Informations de commande: On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Informations de livraison: Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.