Imprimer
Liquid pump sampling tool with glass bottle
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5021G-500 5021X-500
300-0464EA 496 EUR
300-0464 SAMP5021X-500
Liquid pump sampling tool with glass bottle
Échantillonneurs Préleveurs de liquides
This pump sampler set is ideal for contamination-free liquid sampling.
  • Supplied in a convenient carry case
  • Glass bottle container offers excellent chemical resistance
  • Easy to use
  • Suitable for low to medium viscosity liquids
The liquid pump sampler is ideal for sampling liquids from drums and barrels, and is supplied as a set in a handy case. The kit is designed to sample liquids such as water and oil, and is extremely easy to use.
Informations de livraison: Set includes a plastic hand pump, four (4) 500 ml glass bottles, a 13 m LDPE hose, a tube cutter, and carrying case.