Imprimer
AccuPod® Sample thief manual samplers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1360A-301 1360A-303 1360A-601 1360A-1001 1360A-1501 1360A-2001 1360A-603 1360A-1003 1360A-1503 1360A-2003
300-0433EA 2190 EUR
300-0433 300-0434 300-0435 300-0436 300-0437 300-0438 300-0439 300-0440 300-0441 300-0442
AccuPod® Sample thief manual samplers
Échantillonneurs Préleveurs de solides
The AccuPod® Sample Thief is ideal for unit dose sampling of powders and granules.
  • Perfect for unit dose sampling
  • Next generation sampling technology
  • Combines a single-use disposable pod with a reusable stainless steel sampler
  • Designed to minimise sampling errors, when transfering from one container to another
The AccuPod® represents the next generation in sampling technology and combines a single-use disposable pod with a stainless steel sampler for unit dose sampling of powders and granules. This revolutionary sampler allows the sample to stay in the pod until it is needed in the laboratory, therefore reducing sampling errors. It is frequently used in sampling blend uniformity, drug development, and general quality control work.