Imprimer
VWR® AnalaR NORMAPUR®, Glass Wool, Superfine (~11 µm)
Fournisseur: VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
85774.140EA 28.8 EUR
85774.140 85774.180 85774.290
VWR® AnalaR NORMAPUR®, Glass Wool, Superfine (~11 µm)
Glass and Cotton Wool
Glass wool is often used as filter material in analytical processes and as supporting substance for drying agents in drying towers (e.g. to prevent “caking” in P2O5).
  • Chemical resistance even against strong acids (except HF)
Other applications are based on its thermal resistance and its very good insulation properties (up to ~500 °C).