Imprimer
Ball / flange connections, DURAN®
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
01250015 01250025 01260015 01260025
LENZ01250015EA 42.3 EUR
LENZ01250015 LENZ01250025 LENZ01260015 LENZ01260025
Ball / flange connections, DURAN®
Adaptateurs Adaptateurs rodés sphériques
Made from DURAN® tubing.
  • High temperature resistance
  • Very good chemical resistance
Manufactured to inch dimensions in accordance with DIN 12244 and DIN 12264.