Imprimer
MPU series couplings
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MPU171239 MPU221239M
CPCCMPU171239EA 261 EUR
CPCCMPU171239 CPCCMPU221239M
MPU series couplings
Raccords et connecteurs
MPU series twist-to-connect design features an easy-to-use locking mechanism that guards against accidental disconnects and provides a reliable, secure connection.
  • Easy to operate
  • A ³/₄" hose barb provides smooth, rapid media transfer
  • Locking feature guards against accidental disconnects
  • Sharp barb end minimises fluid turbulence and dead space
Shrouded, leak-free seal and smooth, internal protects valuable fluids and eliminate flow path potential to contaminate fluid path.
Operating temperature: −40 to 149 °C (for polysulphone)
Certifications: USP Class VI, ADCF certified and meets BSE/TSE requirements.