Imprimer
AseptiQuik® C connectors
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AQC22008 AQC33012HT AQC44012HT
CPCCAQC22008EA 403 EUR
CPCCAQC22008 CPCCAQC33012HT CPCCAQC44012HT
AseptiQuik® C connectors
Raccords et connecteurs
AseptiQuik® C Connectors provide quick and easy sterile connections, even in non-sterile environments. The 'Click-Pull-Twist' design enables users to transfer media easily with less risk of operator error.
  • Operating temperature: 4 to 40 °C
  • Ensure simultaneous and secure removal of both membranes
  • Audible confirmation of completed assembly steps
  • Open access through multiple supply chain partners
  • Secures final connection preventing disassembly
Biopharmaceutical manufacturers can make sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Certifications: USP class VI, ADCF certified.