Imprimer
MPC series ¹/₄" flow, quick disconnect coupling for bio and pharma
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MPC30003M MPC30039M MPC32003 MPC32003B25 MPC32039 MPC32039B25 MPCK17004T39 MPCK17004T39B2 MPCK17006T03 MPCK17006T03B2 MPCK17006T39 MPCK17006T39B2 MPCK32003 MPCK32003B25 MPCK32039
212-0824EA 1020 EUR
212-0824 212-0825 212-0826 212-0827 212-0828 212-0829 CPCCMPCK17004T39 CPCCMPCK17004T39B2 CPCCMPCK17006T03 CPCCMPCK17006T03B2 CPCCMPCK17006T39 CPCCMPCK17006T39B2 212-0830 212-0831 212-0832
MPC series ¹/₄" flow, quick disconnect coupling for bio and pharma
Raccords et connecteurs
These ¹/₄" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets.
Materials of construction include animal free (BSE/TSE) USP class 6 polysulphone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Informations de commande: Accessories not included