Imprimer
Sanitary fitting to hose barb fitting adapter
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SQCC221212M SQCC222424M SQCX221212M SQCX222424M
212-0815EA 1540 EUR
212-0815 212-0816 212-0817 212-0818
Sanitary fitting to hose barb fitting adapter
Raccords et connecteurs
Stainless steel adapter engineered to ease the transition from stainless steel process pipe and equipment to single use disposable plastic.
Materials of construction include 316L stainless steel and platinum cured, USP class 6 silicone oring seals.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Informations de commande: Accessories not included