Imprimer
Mixed cellulose ester membranes, sterile, 0,2 µm, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7187-114 10401770 10401772 10406970 10406972 10408714 10408712
514-0106EA 185 EUR
514-0106 514-2662 514-2663 514-3363 514-3373 514-1156 511-0138
Mixed cellulose ester membranes, sterile, 0,2 µm, Whatman™
Filtres Filtres à membrane
Whatman sterile MCE membrane filters from Cytiva's business are plain or gridded filters designed to support routine cell counting and particle detection.
  • Excellent contrast for easy particle detection
  • High internal surface area for sample adsorption
  • High dirt loading capacity
  • Biologically inert with good thermal stability
  • Uniform microporous structure delivers high flow rates
These MCE membrane filters are composed of cellulose acetate and cellulose nitrate, giving the membrane a more uniform surface than pure nitrocellulose filters. Other sterile mixed cellulose ester membranes are available.
The MCE membrane filters are biologically inert and possess a number of features that enable contaminant-free cell counting. They are manufactured using a nontoxic ink and without surfactants, decreasing the risk of inhibiting bacterial growth for culturing to determine microbial contamination in microbiological quality control.
MCE membrane filters are available plain for automated colony counting methods, or gridded for manual counting. Grid lines in white, black, or green are spaced at 3,1 mm intervals. The special ink used to apply grids is nontoxic and free from bacterial growth inhibitors.