Imprimer
Sterile cellulose MicroPlus membrane filters, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10407112 10407114 10407132 10407134 10407170 10407172 10407713 10407714 10407734
514-1142EA 1010 EUR
514-1142 514-1143 514-1144 514-1145 514-1146 514-1147 514-1153 514-1154 514-0084
Sterile cellulose MicroPlus membrane filters, Whatman™
Filtres Filtres à membrane
Cytiva's sterile Whatman MicroPlus-21 cellulose nitrate membrane filters are for microbiological quality control. The STL version is compatible with Membrane-Butler and other membrane filter dispensers.
  • All MicroPlus-21 membrane filters are white with a high-contrast black grid (3.1 mm)
  • Designed specifically for microbiological QC
  • High mechanical strength and flow rate
  • Tested for bubble point, bacterial retention, flow rate, recovery rate, and sterility
  • Individually packaged for convenience and sterility
  • Type STL provided in dispenser boxes with 100
En savoir plus