Imprimer
Rubans vinyle à usage général 764
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
764IYL50IW 764IW50IW 764IR50IW 764IBL50IW 764IO50IW 764IBK50IW 764IG50IW
MMMA764IYL50IWEA 179 EUR
MMMA764IYL50IW MMMA764IW50IW MMMA764IR50IW MMMA764IBL50IW MMMA764IO50IW MMMA764IBK50IW MMMA764IG50IW
Rubans vinyle à usage général 764
Rubans adhésifs
Le ruban en vinyle à usage général 3M™ 764 est utilisé pour toutes les applications non critiques, y compris le codage couleur, le regroupement et le marquage de sécurité.
  • Use for taping in all non-critical applications
  • Protects parts, equipment, and products from abrasions
  • Bundle items and temporarily mark parts, machines, and more
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
Find dozens of ways to use 3M™ General-Purpose Vinyl Tape 764. It is non-corrosive, flexible, abrasion-resistant, conformable, wear-resistant, easy to apply, versatile, and moisture-resistant. This flexible vinyl tape is ideal for protecting parts, equipment, and products from abrasions.