Imprimer

Anti-STING mouse monoclonal antibody [clone: 8E7]

Fournisseur: Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

CAYM25922-100EA 575 EUR
CAYM25922-100
Anti-STING mouse monoclonal antibody [clone: 8E7]
Anticorps
Type: Primary
Antigen: STING M284 variant
Clonality: Monoclonal
Clone: 8E7
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR