Imprimer
VWR®, HF Bottle Top Dispensers
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-6863EA 829 EUR
612-6863 612-6864
VWR®, HF Bottle Top Dispensers
Distributeurs pour flacons
Designed with ergonomics and intuitive handling in mind, HF bottle top dispenser helps in dispensing Hydrofluoric Acid with safety and precision.
  • Springless Valve™ for smooth and jam-free functioning
  • PTFE piston with ETP O-Ring for effortless piston movement and high chemical compatibility
  • Easy and effortless volume setting
  • Flexible delivery nozzle to change angle of dispensing
  • Dispensers is fully autoclavable at 121 °C and 15 psi for a duration of 10 to 15 minutes
  • Reproducibility of 0,2%
HF bottle top dispenser is designed with selected materials for barrel and valve assembly that enable the dispenser to be used with Hydrofluoric Acid (HF) in addition to solvents and other reagents.
The VWR® E-BURETTE has a ‘recirculation mode’ which prevents loss of reagents during purging and facilitates bubble-free dispensing. The delivery nozzle of the burette can be adjusted in both horizontal and vertical directions providing easy dispensing in demanding laboratory conditions.
The user-friendly TFT touchscreen guides quick and accurate titrations and displays numeric data up to the second decimal. A hand held stylus is provided as an accessory to assist touch operations on the control panel. The touch screen control panel can be used to perform functions like automatic re-fill without change of reading, zero reset, fill and dispense. The control panel is powered by a rechargeable, long lasting Li-ion battery. This battery can be easily replaced by the user.
The instrument has computer connectivity through a mini USB cable. It has a built-in software which allows readings to be transferred and saved in a file on the computer.
Certifications: Meets ISO 8655 standard for accuracy and precision, manufacturing standards: ISO 9001, ISO 13485, ISO 17025 and product certification: CE
Informations de commande: Supplied with a digital bottle top burette, a control panel, a micro-USB cable, telscoping tube, five thread adapters (28, 32, 38, 40 and 45 mm), stylus, amber colored window, user instruction manual and an individual quality control certificate that documents the serial number and test values, and confirms the conformity according to weights and measures regulations as per ISO 8655.
Attention: Maintain instrument and reagent temperature between +10 to +40 °C (From 50 to 104 °F). Maintain reagent vapor pressure up to 600 mbar (maximum). Aspirate slowly above 300 mbar pressure in order to prevent the liquid from boiling. Kinematic viscosity up to 500 mm²/s. Relative air humidity between 20 and 90%. Use fluids with density up to 2,2 g/cm³.