Imprimer
dNTP mix
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40053 40054
40053.EA 250 EUR
40053. 40054.
dNTP mix
Réactifs pour acides nucléiques Nucléotides
Highly purified mix of dNTPs (>99% by HPLC) in 0,6 mM Tris-HCl (pH 7,5); each dNTP is at a concentration of 10 or 25 mm in the solution provided.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.